This item is now discontinued. Please browse similar items
Corning 430658 Cryogenic Vials, External Thread with Silicone Washer, Conical Self-Standing, 1.2 mL; 50/Pk
Representative image only.
Corning Mfr # 430658 – Item # WZ-44351-00
Safely and securely store your cryogenic samples
  • Supplied sterile and certified non-pyrogenic
  • Polypropylene construction withstands temperatures to -196°C
  • Silicone washers provide a secure seal
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)1.2
  • Tube MaterialPolypropylene
  • Tube Bottom ShapeConical self-standing
  • Tube ColorNatural
  • Cap TypeScrew
  • Cap MaterialPolypropylene
  • Cap colorOrange
  • ThreadExternal
  • SterileYes
  • DescriptionCryogenic Vials, External Thread with Silicone Washer, Conical Self-Standing, 1.2 mL; 50/Pk
Key Features
  • Supplied sterile and certified non-pyrogenic
  • Polypropylene construction withstands temperatures to -196°C
  • Silicone washers provide a secure seal
More About this Item
Vials feature a self-standing design with black graduations and larger marking spots for easier filling and sample identification. Choose from internal or external threaded vials for a wide range of applications.
Vial caps will hold Corning® color-coded inserts and will lock into Corning® cryogenic racks for single-handed operation. Cap inserts and self-locking racks are sold separately.