This item is now discontinued. Please browse similar items
Corning 3262 ultra low attachment surface culture dish, round dish, 100 mm dia
Representative image only.
Corning Mfr # 3262 – Item # WZ-01835-22
Unique hydrogel surface inhibits cell attachment
SPECS & DESCRIPTION
Specifications & Description
  • Well Diameter (mm)100
  • Surface TreatmentHydrogel
  • Descriptionultra low attachment surface culture dish, round dish, 100 mm dia
Nonpyrogenic
More About this Item
These culture plates and dishes have a unique covalently bonded hydrogel surface that is hydrophilc and neutrally charged. It minimizes cell attachment, protein absorption and enzyme activation. The surface is non-cytotoxic, biologically inert and nondegradable. Use these products to maintain cells in a suspended, unattached state, prevent stem cells from attachment-mediated differentiation, prevent anchorage-dependent cells from dividing, and reduce binding of attachment and serum proteins to the substrate.