Cole-Parmer Essentials Cork Borer, 9 Piece Set, Plated Brass
Representative image only.
Cole-Parmer Essentials Item # WZ-06298-98
Easily convert solid stopper into holed stopper
  • Steel punch makes boring easier
  • 1 year warranty
PRODUCT OPTIONS
£65.31GBP / Each
£78.37GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass
  • DescriptionCork Borer, 9 Piece Set, Plated Brass
  • Warranty1 year
1 Year Warranty
Key Features
  • Steel punch makes boring easier
  • 1 year warranty
More About this Item
Convert your solid stopper into holed-stopper with these easy-to-use borer sets. All borers come with a steel punch to make boring easier. Borer sharpener (sold separately as part number 06298-94) keeps your borer in top condition and sharpens borers up to 25mm in diameter.
Boring diameter 4 to 15 mm