This item is now discontinued. Please browse similar items
CoorsTek 60383 Contoured Alumina Pestle, unglazed grinding surface
Representative image only.
CoorsTek Mfr # 60383 – Item # WZ-17822-34
SPECS & DESCRIPTION
Specifications & Description
  • Pestles Length (mm)110
  • DescriptionContoured Alumina Pestle, unglazed grinding surface
More About this Item
Contoured shape of mortar and pestle allows easy holding. High alumina composition is extremely hard and resistant to abrasion. Mortar is glazed except for outside bottom and inside grinding surface; pestle is glazed except for bottom grinding surface. Mortar and pestle are sold separately. Autoclavable.