This item is now discontinued. Please browse similar items
Cooper-Atkins 4005I Cooper/Atkins Thermometer, 1/4-3/8" Pipe Clamp and 2.5" Penetration Probe
Representative image only.
Cooper-Atkins Mfr # 4005I – Item # WZ-90025-03
Easy single handed temperature measurement
  • Spring-loaded cordless pipe clamp ensure repeatability
  • 2.5” retractable general purpose probe
  • Single-handed use
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermistor
  • Number Of Probe Channels1
  • Types Of Probes AcceptedThermistor
  • Min Temperature (° F)-20
  • Max Temperature (° F)300
  • Min Temperature (° C)-29
  • Max Temperature (° C)150
  • Accuracy±3.0°F (±2.0°C)
  • Resolution0.1°
  • Display Type0.25\" Display, 3-digit LCD
  • No. of Probe(s) Included1
  • Probe Length (in)2 1/2
  • Battery Life90 hours
  • Battery1.5v Battery (1)
  • DescriptionCooper/Atkins Thermometer, 1/4-3/8\" Pipe Clamp and 2.5\" Penetration Probe
  • Warranty1 year
1 Year Warranty
Key Features
  • Spring-loaded cordless pipe clamp ensure repeatability
  • 2.5” retractable general purpose probe
  • Single-handed use
  • 1 year warranty
More About this Item
This versatile instrument measures the surface temperature of a pipe and other round objects or the internal temperature of liquids and solids. The claw like spring loaded clamp probe is designed to accommodate round objects from ¼" to 1-3/8" in diameter. The retractable stainless steel penetration probe is hidden in the handle of the unit and can be inserted in to an object up to 2" deep. Temperature is conveniently shown on a built-in LCD. Other features include auto shut-off, unit of measure, min/max and hold.