Cole-Parmer WP5BB97P Fibrous polypropylene string-wound cartridge, 5 µm, 10" length
Representative image only.
Cole-Parmer Item # WZ-01508-76
String-wound pattern provides greater surface area for lower pressure drop and higher flow
PRODUCT OPTIONS
£25.66GBP / Each
£30.79GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating5
  • Filter Length (in)10
  • DescriptionFibrous polypropylene string-wound cartridge, 5 µm, 10" length
More About this Item
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
Fibrous polypropylene cartridges have a lower pressure drop than fibrillated cartridges but use an antistatic agent that may cause foaming in some applications.