This item is now discontinued. Please browse similar items

Cole-Parmer Vertical Dual Adjustable Slab Gel System, CE-marked

Representative image only.
Cole-Parmer Item # WZ-28570-15
Customize your own gel system depending on your needs and budget!
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)900
  • Cell Width (in)7.5
  • Cell Depth (in)9.5
  • Cell Width (cm)19.05
  • Cell Height (cm)31.12
  • Cell Depth (cm)24.13
  • Glass Width (in)6.5
  • Glass Length (in)11.02
  • Glass Width (cm)16.5
  • Glass Length (cm)28
  • DescriptionVertical Dual Adjustable Slab Gel System, CE-marked
  • Warranty3 year
3 Year Warranty

Key Features

  • 3 year warranty

More About this Item

Versatile units are adjustable for different pathlengths. Ideal for SDS page, sequencing, mobility shift assays, and characterizing or separating proteins. The upper reservoir is height adjustable along the guide rods, and creates a positive, leakproof seal when plate assembly is clamped into place.

Required System Components:
1) Gel Unit (order power supply separately)

For the Gel Wrap™ Gasket gel casting method, you will need:
2) Glass plate set(s)
3) Casting clamps (six per plate set for height-adjustable units)

Order the same thickness of:
4) GelWrap gasket (one per plate set)
5) Spacer set (one per plate set)
6) Combs (one per plate set)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat