Cole-Parmer VapLock™ Fitting, Yellow PP w/ Orange ETFE, Straight, Compression to Threaded Adapter, 3.0 mm OD x 1/4-28 UNF(M); 10/PK
Representative image only.
Cole-Parmer Item # WZ-12020-07
Designed for secure liquid-tight connections
  • Two piece nut and ferrule system
  • Adapter for semi-rigid tubing
  • Color coded for your convenience
PRODUCT OPTIONS
£58.71GBP / Pkg of 10
£70.45GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPP / ETFE
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (in)1/4-28
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)3
  • ColorYellow / Orange
  • DescriptionTubing Adapter, 3.0 mm OD x 1/4-28 UNF(M), yellow PP w/ orange ETFE; 10/PK
Key Features
  • Two piece nut and ferrule system
  • Adapter for semi-rigid tubing
  • Color coded for your convenience
More About this Item
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. A two-piece nut and ferrule system provides a liquid-tight connection and withstands pressures up to 500 psi.