This item is now discontinued. Please browse similar items
Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Compression to Threaded Adapter, 5/8" OD Tubing x 1/2" NPT(M); 5/PK
Representative image only.
Cole-Parmer Item # WZ-12020-42
Designed for secure leak-resistant connections
  • Constructed of durable polypropylene
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeCompression x NPT(M)
  • Thread size NPT(M) (port 2) (in)1/2
  • Compression Size / Tube Od (Port 1) (in)5/8
  • Compression Size / Tube Od (Port 1) (mm)15.9
  • DescriptionFitting, Polypropylene, Straight, Compression to Threaded Adapter, 5/8\" OD Tubing x 1/2\" NPT(M); 5/PK
Key Features
  • Constructed of durable polypropylene
More About this Item
Adapters with large bore for semi-rigid tubing are constructed of two-piece molded polypropylene components and do not include a separate ferrule. Adapters withstand pressures up to 100 psi. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.