Cole-Parmer VapLock™ Luer Fitting, Polypropylene, Straight, Male Luer Lock x 1/16" Hose Barb; 10/Pk
Representative image only.
Cole-Parmer Item # WZ-12028-47
Quick connection and release make these luer fittings excellent for many applications
  • Great for connecting to syringes and small tubing
PRODUCT OPTIONS
£9.51GBP / Pkg of 10
£11.41GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene (PP)
  • Number of Ports2
  • Connection Type-Port 1Straight male luer lock
  • Connection Type-Port 21/16\" hose barb
  • DescriptionLuer Fitting, PP, Straight, Male Luer Lock x 1/16\" Hose Barb; 10/PK
Key Features
  • Great for connecting to syringes and small tubing
More About this Item
Choose from fittings constructed from durable polypropylene (PP) or chemically inert PVDF.