Cole-Parmer VapLock™ Adapters for Exhaust Filters, Justrite® Centura Quick-Disconnect, 90° Elbow, Stainless Steel and PTFE
Representative image only.
Cole-Parmer Item # WZ-12019-92
Various adapter configurations allow quick and easy connection to emission sources
  • Suitable for use with Justrite® Centura safety containers
PRODUCT OPTIONS
£63.66GBP / Each
£76.39GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless steel/PTFE
  • O-Ring MaterialPTFE
  • Connection TypeNPT(M) x Quick Disconnect, 90° elbow
  • Thread size NPT(M) (port 2) (in)1/4
  • ColorGray
  • DescriptionFilter Adapter, 90° elbow, Justrite Centura quick disconnect, SS and PTFE; 1/ea
Key Features
  • Suitable for use with Justrite® Centura safety containers
More About this Item
VapLock exhaust filters have a 1/4" NPT(F) port and may require an adapter to work with your container. Quick-disconnect adapters fit Justrite® Centura safety containers.