This item is now discontinued. Please browse similar items
Cole-Parmer UN/DOT Waste System Cap w/ Filter, 51 mm, 6 x 1/16" OD tubing
Representative image only.
Cole-Parmer Item # WZ-06062-01
Collect hazardous waste safely
  • UN/DOT approved cap suitable for packing groups II and III
  • Activated carbon exhaust filter traps solvent vapors
  • Included elbow adapter saves vertical space
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPolypropylene
  • Cap / Stopper Size51 mm
  • Cap ColorWhite
  • Special Features6 x 1/8\" OD tubing
  • DescriptionUN/DOT Waste System Cap w/ Filter, 51 mm, 6 x 1/8\" OD tubing
Key Features
  • UN/DOT approved cap suitable for packing groups II and III
  • Activated carbon exhaust filter traps solvent vapors
  • Included elbow adapter saves vertical space
More About this Item
When paired with a UN/DOT-approved container with the same rating*, these carboy cap assemblies create a safe system for chemical waste collection and disposal. Fittings secure waste lines to cap, providing a closed loop to prevent harmful fumes from escaping. Activated carbon exhaust filter adsorbs vapors, allowing only clean air to return to the lab environment. Polypropylene cap with sulfur-cured EPDM gasket and fittings made of acetal and ETFE are chemically resistant to most HPLC-grade solvents.