Cole-Parmer Tuff-Tip, Semi-Dome 1" In-Line or Submersible pH Electrode, BNC
Representative image only.
Cole-Parmer Item # WZ-27020-26
Less chance of breakage
  • Choose chemical-resistant Ryton® PPS or graphite-tipped electrodes
  • 1 year warranty
PRODUCT OPTIONS
£267.00GBP / Each
£320.40GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Electrode Body MaterialKynar®
  • Reference JunctionCeramic
  • Max Temperature (° C)110
  • Temperature CompensationYes
  • ATC ConnectorSpade lugs
  • ATC Element1 kΩ Pt RTD
  • ConnectorBNC
  • DescriptionTuff-Tip, Semi-Dome 1\" In-Line or Submersible pH Electrode, BNC
  • Warranty1 year
1 Year WarrantyKeep a Spare
Key Features
  • Choose chemical-resistant Ryton® PPS or graphite-tipped electrodes
  • 1 year warranty
More About this Item
Don't worry about bulb breakage— unique design helps protect bulbs
– Eliminate electrical noise with graphite-tipped models
– Ideal for viscous or severe applications
– Reduce contamination and clogging with uniform pore size