This item is now discontinued. Please browse similar items
Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/Tinned
Representative image only.
Cole-Parmer Item # WZ-05995-50
Semi-domed, tuff-tip limits glass exposure and protects electrode from damage
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialPDVF
  • Min pH (pH)0
  • Max pH (pH)13
  • Min Temperature (° C)-5
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • GuardTuff-Tip
  • Thread Size NPT(M) (in)1/2
  • ATC ConnectorSpade lugs
  • ATC Element1 kΩ Pt RTD
  • ConnectorTinned ends
  • Cable Length (m)3
  • DescriptionTuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/Tinned
  • Warranty1 year
1 Year Warranty
Key Features
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
  • 1 year warranty
More About this Item
Electrodes feature PVDF housing with 1/2'' NPT(M) threads and withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Electrodes include 10-ft cable with BNC pH connector and tinned ends for ATC element. Insertion depth 1''.