This item is now discontinued. Please browse similar items
Cole-Parmer Trinocular Stereozoom Microscope, 10x-40x; 230V
Representative image only.
Cole-Parmer Item # WZ-48920-25
Stereozoom optics let you easily focus through different magnification ranges
  • Adjustable illumination for all your lighting needs
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification10x to 40x
  • Maximum Magnification40x
  • Head ConfigurationTrinocular
  • Objectives1x to 4x
  • Eyepiece Magnification10x
  • EyepiecesWidefield
  • Illuminator(s)Top and bottom
  • Stand TypeStandard
  • Power (VAC)230
  • DescriptionTrinocular Stereozoom Microscope, 10x-40x; 230V
  • WarrantyLTD Life Time
Conformite EuropeenneLimited Lifetime Warranty
Key Features
  • Adjustable illumination for all your lighting needs
  • Limited Lifetime Warranty
More About this Item
Stereozoom microscopes feature stepless magnification through zoom control on viewing head. Head rotates 360°; raise or lower the head for viewing different size objets. Widefield 10x eyepieces feature adjustable diopter and interpupillary distance for comfortable viewing.
Standard stand provides built-in incident (top) and transmitted (bottom) illumination; intensity is adjustable through control on side of base. Also includes a reversible black/white stage plate.
Use the trinocular port for mounting a 35-mm camera or video camera for your photomicroscopy applications. Trinocular microscopes include a video adapter with a 0.4x lens, and an SLR adapter with 2.0x lens; T-mount not included, order separately.
includes a 6-ft cord and a European plug.