Cole-Parmer Essentials Trace Metal Analysis Pipette Tips, Non-Sterile, 101 to 1000 µL; 10 Racks of 100 Tips
Representative image only.
Cole-Parmer Essentials Item # WZ-25713-00
Tips are certified trace-metal free to prevent sample contamination
  • Constructed of virgin polypropylene
  • All materials used in the manufacture of these tips are free of trace metals
  • Packaged as 100 Tip Rack with Lift-off Lid
PRODUCT OPTIONS
£135.67GBP / Pkg of 1000
£162.80GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)1000
  • Min Volume (µL)101
  • Tip Volume (mL)1
  • Overall Length (in)2
  • Overall Length (mm)50.8
  • Tip StyleTrace Metal Free
  • Wetted MatterialPolypropylene
  • Color CodeNatural
  • Package Of10 Racks of 100 Tips
  • Compatible withCorning; Costar®; Eppendorf®; Finnpipette®; Hamilton; Nichiryo; Oxford® BenchMate™; Pipetman®; Rainin EDP2; Titertek®; Socorex
  • DescriptionTrace Metal Analysis Pipette Tips, Non-Sterile, 101 to 1000 µL; 10 Racks of 100 Tips
Key Features
  • Constructed of virgin polypropylene
  • All materials used in the manufacture of these tips are free of trace metals
  • Packaged as 100 Tip Rack with Lift-off Lid
More About this Item
These universal pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit to variety of pipettors to ensure laboratory standardization. A reference mark at 10 µL lets you check pipettor calibration and visually estimate sample size. Tips are packaged in racks for easy access. Tips are tested by an independent laboratory and certified to be RNAse- and DNAse- free, a trace metal certificate is available upon request.