Cole-Parmer Essentials Testing Sieve, 8" OD Brass Frame/Brass Wire, Half Height, No. 325
Representative image only.
Cole-Parmer Essentials Item # WZ-59981-23
Get more accurate results
  • Sievers are serialized
  • Come with a test sieve certificate of compliance
  • 1 year warranty
PRODUCT OPTIONS
£215.16GBP / Each
£258.19GBP (incl VAT)
£126.63 - £185.17GBP / Each
£151.96 - £222.20GBP (incl VAT)
for an additional
£126.63GBP / Each
£151.96GBP (incl VAT)
for an additional
£185.17GBP / Each
£222.20GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass frame, brass wire
  • U.S. Standard Screen Size (No.)325
  • Tyler Standard Screen Size (mesh)325
  • Sieve Opening (mm)0.044
  • Outside Diameter (in)8
  • Height (InchShort)1
  • DescriptionTesting Sieve, 8\" OD Brass Frame/Brass Wire, Half height, No. 325
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Sievers are serialized
  • Come with a test sieve certificate of compliance
  • 1 year warranty
More About this Item
These brass testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.