This item is now discontinued. Please browse similar items
Cole-Parmer Wireless Terminal Interface; 1/Each
Representative image only.
Cole-Parmer Item # WZ-10100-95
  • Allows for wireless communication between the balance and control terminal
  • Lasts up to 8 hours on a single change
  • Ten m maximum range—have your balance under a fume hood and control panel outside
  • Obtain more accurate readings
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeWireless Terminal
  • DescriptionWireless Terminal Interface; 1/Each
1 Month Warranty
Key Features
  • Allows for wireless communication between the balance and control terminal
  • Lasts up to 8 hours on a single change
  • Ten m maximum range—have your balance under a fume hood and control panel outside
  • Obtain more accurate readings
  • 1 month warranty
More About this Item
Replacement parts and accessories for your Cole-Parmer Basic and Advanced Touch-Screen Balances.
Since the terminal doesn't have to share benchspace with the weighing module, additional vibrations are elimated—providing more accurate and stable measurements.