This item is now discontinued. Please browse similar items

Cole-Parmer Submarine Horizontal Gel System

Representative image only.
Cole-Parmer Item # WZ-28561-00
Cooling chamber and buffer circulation ports let you maintain buffer temperature and pH during long runs
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)1500
  • Gel Width (cm)14
  • Gel Length (cm)20
  • Cell Width (in)13.75
  • Cell Height (in)6
  • Cell Depth (in)4
  • Cell Width (cm)5.41
  • Cell Height (cm)15.24
  • Cell Depth (cm)1.57
  • DescriptionSubmarine Horizontal Gel System
  • Warranty3 Years
3 Year Warranty

Key Features

  • 3 year warranty

More About this Item

Multiple comb slots let you configure gels for single runs or multiple simultaneous short runs — ideal for screening large numbers of samples. Buffer circulation ports let you maintain uniform pH and ionic strength. Cooling chamber allows flow of water under gel bed to maintain constant temperature. Tapered-baffle design lets you cast gels directly in the unit — no sealing or taping needed. UV-transparent acrylic gel tray has ''anchors'' to prevent gel from floating during electrophoresis. Base of unit has built-in leveling feet.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat