This item is now discontinued. Please browse similar items
Cole-Parmer Sterile PS Culture Tubes w/ 2-Position PE Caps; 12x75mm, Clear; 1000/Bx
Representative image only.
Cole-Parmer Item # WZ-06339-95
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPE
  • Length (mm)75
  • Depth (mm)75
  • Diameter (mm)12
  • DescriptionSterile PS Culture Tubes w/ 2-Position PE Caps; 12x75mm, Clear; 1000/Bx
  • Warranty1 year
1 Year WarrantySterile
Key Features
  • 1 year warranty
More About this Item
Heavy-walled tubes resist breakage. Choose from clear polystyrene (PS) or natural polypropylene (PP). The 12 x 75 mm tubes have a volume of 6 mL; the 17 x 100 mm tubes have a volume of 16 mL. Tubes are sterile; include two-position PE caps.