Cole-Parmer Sterile Petri Dishes; 60 mm dia x 15 mm H; 500/box

Representative image only.
Cole-Parmer Item # WZ-06139-01

£127.05GBP / of 500
£152.46GBP (incl VAT)
Parafilm PM992 Wrap , 2
£25.05GBP / Roll of 1
£30.06GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolystyrene
  • Overall Height (in)9/16
  • Overall Height (cm)1.5
  • Outside Diameter (OD) (in)2
  • Outside Diameter (OD) (cm)6
  • DescriptionSterile Petri Dishes; 60 mm dia x 15 mm H; 500/box

More About this Item

Disposable polystyrene dishes have convenient stacking ridges. Packed sterile in easy-open bags of 20. Gamma irradiated. 2 bx/cs*.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat