This item is now discontinued. Please browse similar items
Cole-Parmer StableTemp Stainless Steel Sand Baths, 1 Block Models
Representative image only.
Cole-Parmer StableTemp Item # WZ-36600-57
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)2 1/4
  • Width (in)3 3/4
  • Length (in)2 1/2
  • DescriptionStainless Steel Sand Baths, 1 Block Models
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Stainless Steel Sand Baths are ideal for irregular vessels or to hold a variety of different size vessels at one time. Constructed of stainless steel for superior corrosion resistance. Designed to hold sand, stainless steel shot, or non-volatile liquids. Fits into the Cole Parmer Stable Temp Dry Block Heaters in place of a modular block. Select the appropriate capacity for the size block heater.
Stainless Steel Sand Baths are ideal for irregular vessels or to hold a variety of different size vessels at one time. Constructed of stainless steel for superior corrosion resistance. Designed to hold sand, stainless steel shot, or non-volatile liquids. Fits into the Cole Parmer Stable Temp Dry Block Heater one block capacity models in place of a modular block.