Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 7.5; 13/Pk
Representative image only.
Cole-Parmer Essentials Item # WZ-62991-24
£64.65GBP / Pkg of 13
£77.58GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number7.5
  • Top Outside Diameter (OD) (mm)39
  • Bottom Outside Diameter (OD) (mm)31
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 7.5; 13/Pk
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.