Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 16; 1/Pk
Representative image only.
Cole-Parmer Essentials Item # WZ-62991-52
£101.97GBP / Pkg of 1
£122.36GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number16
  • Top Outside Diameter (OD) (mm)127
  • Bottom Outside Diameter (OD) (mm)90
  • Height (mm)50
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 16; 1/Pk
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.