Cole-Parmer Essentials Solid Green Neoprene Stoppers, Standard Size 13.5; 3/Pk
Representative image only.
Cole-Parmer Essentials Item # WZ-62991-46
£96.54GBP / Pkg of 3
£115.85GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number13.5
  • Top Outside Diameter (OD) (mm)75
  • Bottom Outside Diameter (OD) (mm)62
  • Height (mm)35
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 13.5; 3/Pk
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.