This item is now discontinued. Please browse similar items
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 11.5; 4/Pk
Representative image only.
Cole-Parmer Item # WZ-62991-40
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number11.5
  • Top Outside Diameter (OD) (mm)63
  • Bottom Outside Diameter (OD) (mm)50
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 11.5; 4/Pk
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.