Cole-Parmer Essentials Solid Black Rubber Stoppers, Standard Size 14; 1/Pk
Representative image only.
Cole-Parmer Essentials Item # WZ-62990-48
£48.51GBP / Pkg of 1
£58.21GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number14
  • Top Outside Diameter (OD) (mm)90
  • Bottom Outside Diameter (OD) (mm)75
  • Height (mm)39
  • ColorBlack
  • MaterialRubber
  • DescriptionSolid Black Rubber Stoppers, Standard Size 14; 1/Pk
More About this Item
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.