Cole-Parmer Software for Single-Channel Flame Photometers
Representative image only.
Cole-Parmer Item # WZ-02656-15
Cature results automatically and write reports
£909.75GBP / Each
£1,091.70GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSoftware for Single-Channel Flame Photometers
More About this Item
The optional software allows you to add names/sample numbers, show visual indication, do multi-point calibration, report writing, and capture results automatically. Requires a PC running Windows XP. The software, when used with the digital interface, also allows you to use the Autosampler (02657-25) with the single-channel flame photometer.