This item is now discontinued. Please browse similar items
Cole-Parmer Single-Use Sterile Media Bottle, PETG, 38-430 Cap, 1000 mL; 12/Pk
Representative image only.
Cole-Parmer Item # WZ-06061-53
Avoid time-consuming preparation with presterilized media bottles
  • Manufactured in a certified ISO Class 7 cleanroom
  • Assemblies are gamma irradiated and made entirely with USP Class VI components
  • Constructed of durable BPA-free PETG
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)1000
  • Cap MaterialPolypropylene
  • Cap / Stopper Size38-430
  • Cap ColorWhite
  • Bottle ColorClear
  • DescriptionSingle-Use Sterile Media Bottle, PETG, 38-430 Cap, 1000 mL; 12/Pk
Sterile
Key Features
  • Manufactured in a certified ISO Class 7 cleanroom
  • Assemblies are gamma irradiated and made entirely with USP Class VI components
  • Constructed of durable BPA-free PETG
More About this Item
These single-use media bottles with closed polypropylene (PP) cap are ideal for storage and single-use applications. These bottles come presterilized and manufactured in an ISO Class 7 cleanroom, making them ideal for food or pharmaceutical applications. The included cap ensures a closed, secure system without any machining residue or bonding agents. Choose from two material as well as five size options, for a wide range of applications.