This item is now discontinued. Please browse similar items
Cole-Parmer Single-stage Regulator, 1000 scfh Capacity, 320 CGA fitting
Representative image only.
Cole-Parmer Item # WZ-98210-30
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Delivery Pressure (psig)100
  • Max Inlet Pressure (PSI)3000
  • Outlet Connections¼\" NPT(M)
  • CGA Fitting320
  • Capacity (SCFH)1000
  • Outlet Gauge Graduations (PSI)5
  • Inlet Gauge Graduations (PSI)100
  • DescriptionSingle-stage Regulator, 1000 scfh Capacity, 320 CGA fitting
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Ideal for corrosive or noncorrosive gas applications. Designed and constructed for diffusion resistance and easy cleanup. Only brass, stainless steel (SS), PTFE, and Kel-F� contact the operating media.
Metal diaphragm, packless valves, and high-purity seals minimize or eliminate outgoing and inboard diffusion. Supplied with SS screen filter (25 µm); and brass body, bonnet, and gauges. Single-Stage High-Purity Regulators are recommended for analytical work where precise pressure control is not critical.