Cole-Parmer Essentials Sieve; No 200, Full-Height, 8", Brass/Brass, NIST-Traceable Calibration Certificate
Representative image only.
Cole-Parmer Essentials Item # WZ-59980-70
Sieve openings are measured with a computerized video imaging system
  • 1 year warranty
PRODUCT OPTIONS
£268.78GBP / Each
£322.54GBP (incl VAT)
£126.63 - £185.17GBP / Each
£151.96 - £222.20GBP (incl VAT)
for an additional
£126.63GBP / Each
£151.96GBP (incl VAT)
for an additional
£185.17GBP / Each
£222.20GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass frame, brass wire
  • U.S. Standard Screen Size (No.)200
  • Tyler Standard Screen Size (mesh)200
  • Sieve Opening (mm)0.074
  • Outside Diameter (in)8
  • Height (InchShort)2
  • DescriptionSieve; No 200, Full-Height, 8\", Brass/Brass, NIST Cal Cert
  • Warranty1 Year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • 1 year warranty
More About this Item
Sieve is serialized, comes with a test sieve certificate, and meets ASTM E11 specifications. Includes an NIST-traceable calibration report supplied by the manufacturer.