Cole-Parmer Essentials Short-Length Serological Pipette, 25 mL, Ind. Pack; 100/CS
Representative image only.
Cole-Parmer Essentials Item # WZ-34567-57
Easily recognize pipette capacity—color-coded stripes and matching packaging
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Premium material barrier plug
  • Material plug protects your pipette controller from biocontamination and harmful vapors⌡Short pipettes are ideal when working in cramped places—like in hoods
PRODUCT OPTIONS
£84.80GBP / Case of 100
£101.76GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)25
  • Accuracy (%)2%
  • Tip StyleStandard
  • Body MaterialPolystyrene
  • Color CodeRed
  • Filter TypeMaterial plug
  • Package Of25 bulk wrapped per bag
  • DescriptionShort-Length Serological Pipette, 25 mL, Ind. Pack; 100/Cs
Sterile
Key Features
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Premium material barrier plug
  • Material plug protects your pipette controller from biocontamination and harmful vapors⌡Short pipettes are ideal when working in cramped places—like in hoods
More About this Item
These sterile serological pipettes feature bold-colored positive and negative graduations making them easy to read while flling or dispensing. Color coded stripes and packaging enable quick volume identification.
All serological pipettes are manufactured to the ASTM Standard Specification for Serological Pipettes E-934.
Short serological pipettes are ideal for compact workspaces—designed with ergonomics in mind