Cole-Parmer Self-Supportng Dual Arm Gooseneck Fiber Optic Light Pipe, 23" (58.42 cm)
Representative image only.
Cole-Parmer Item # WZ-41720-65
No support needed
  • Flexible arms offer positioning versatility
  • Ideal for use with two microscopes or two-sided illumination of a single sample
  • 1 year warranty
PRODUCT OPTIONS
£388.81GBP / Each
£466.57GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Light Pipe Length (in)23
  • Light Pipe Outside Dimension (OD) (in)0.46
  • Light Pipe Length (cm)58.42
  • Light Pipe Outside Dimension (OD) (cm)1.1684
  • DescriptionSelf-Supporting Light Pipe, Fiber Optic Dual
  • Warranty1 Year
1 Year Warranty
Key Features
  • Flexible arms offer positioning versatility
  • Ideal for use with two microscopes or two-sided illumination of a single sample
  • 1 year warranty
More About this Item
Self-supporting light pipes require no stand or additional support. They connect directly to the illuminator. Flexible gooseneck is constructed of glass fibers sheathed in a steel monocoil.
Illumination base (41720-40) sold separately.