This item is now discontinued. Please browse similar items
Cole-Parmer Self-Cleaning Retractable ORP Electrode with PVDF Housing
Representative image only.
Cole-Parmer Item # WZ-27105-50
Insert up to 12" into pressurized pipes or tanks
  • Dual junctions for demanding applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSelf-cleaning
  • Electrode TypeDouble junction
  • Electrode Body MaterialKynar®
  • ConnectorBNC
  • DescriptionSelf-Cleaning Retractable ORP Electrode with Kynar Housing
  • Warranty1 year
1 Year Warranty
Key Features
  • Dual junctions for demanding applications
  • 1 year warranty
More About this Item
These retractable electrodes feature a flat measuring surface to minimize electrode coating and breakage. Fluid flow across the flat surface increases electrode life and performance. Use this electrode with the 1" port ball valve (order separately as 27015-80) to service your electrode without shutting down the process. For specialty applications, select electrodes for low-ionic strength solutions and high-temperature applications. Mount in any position—connection is 1" NPT(M). Electrodes