This item is now discontinued. Please browse similar items
Cole-Parmer SDS-7025 Standard cell refill solution, 250 mL
Representative image only.
Cole-Parmer Item # WZ-27400-75
Accessories for the Cole-Parmer pH and ORP differential sensors
  • Quick and easy maintenance with optional replacement salt bridge
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeStandard cell refill solution
  • MaterialPPS
  • DescriptionStandard cell refill solution, 250 mL
1 Year Warranty
Key Features
  • Quick and easy maintenance with optional replacement salt bridge
More About this Item
Sensor life is extended with the replacement salt bridge feature and refill solution, which avoids having to replace the entire sensor. Choose from the replacement salt bridge in packs of either 1, 3, 10 10 each. The standard cell refill solution is available in three sizes as well.