Cole-Parmer Screw-Top Microcentrifuge Tube Rotor Adapters; 20/Pk
Representative image only.
Cole-Parmer Item # WZ-06245-53
Enhance the performance of your Cole-Parmer microcentrifuge tubes
  • Compatible with skirted test tube
£42.67GBP / Pkg of 20
£51.20GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionScrew-Top Microcentrifuge Tube Rotor Adapters; 20/Pk
Key Features
  • Compatible with skirted test tube
More About this Item
Parts and accessories to meet your application needs.
Adapts your fixed angle rotor to accept your skirted, screw-top microcentrifuge tubes (06245-52).