Cole-Parmer Vial Cap, Screw Thread Closure, Black, Open-Top, 8-425 mm; 100/Pk
Representative image only.
Cole-Parmer Item # WZ-98721-12
Keep your samples safe with these vial accessories
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This cap requires seals 98721-21 or 98721-22 ordered separately
  • Limited Lifetime Warranty
PRODUCT OPTIONS
£7.64GBP / Pkg of 100
£9.17GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeScrew-Thread
  • Cap Diameter (mm)8
  • Cap colorBlack
  • DescriptionScrew Thread Closure, Black, Open-Top, 8-425 mm
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This cap requires seals 98721-21 or 98721-22 ordered separately
  • Limited Lifetime Warranty
More About this Item
Suitable for use with Cole-Parmer® Versi-Vials
Designed for use with all autosamplers. Seals required and sold separately.