Cole-Parmer Screw-Caps for Microcentrifuge Tubes, Red; 1000/Pk
Representative image only.
Cole-Parmer Item # WZ-06245-57
Color-code your samples
PRODUCT OPTIONS
£143.02GBP / Pkg of 1000
£171.62GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene (PP)
  • ColorRed
  • Compatible WithCole-Parmer Screw-Top Microcentrifuge Tubes (06245-50, 06245-51, 06245-52)
  • DescriptionScrew-Caps for Microcentrifuge Tubes, Red; 1000/Pk
More About this Item
Screw-Caps for Cole-Parmer® Screw-Cap Tubes are available in six colors so you can color code tubes to specific sample type, application, or date. Polypropylene caps include O-rings of ethylene propylene to ensure a leakproof seal.