This item is now discontinued. Please browse similar items
Cole-Parmer Removable Single-Junction ORP Electrode, Tinned ends
Representative image only.
Cole-Parmer Item # WZ-27009-36
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • ConnectorBNC
  • DescriptionRemovable Single-Junction ORP Electrode, Tinned ends
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
More About this Item
Use for oxidation/reduction measurements in plating baths, waste-water monitoring, and other general-purpose applications. Platinum band ORP withstands temperatures from 23 to 212ºF (-5 to 100ºC).