This item is now discontinued. Please browse similar items
Cole-Parmer Rack Insert, Full-Size, for 500 mL Bottles
Representative image only.
Cole-Parmer Item # WZ-98951-90
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With98950-01, 98950-11, 98950-20, 98950-30
  • DescriptionRack Insert, Full-Size, for 500 mL Bottles
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Insert fits either the upper or lower general purpose racks included with the Cole-Parmer Glassware Washers. Full size insert will take up an entire rack, and holds up to 20 (500 mL) bottles.