This item is now discontinued. Please browse similar items
Cole-Parmer PTFE Membrane, 0.20 37mm Wp 100/pk
Representative image only.
Advantec Item # WZ-36230-42
Unaffected by even the most corrosive compounds, including concentrated acids, alkaline, and propellants
  • Durable laminated PTFE membranes
  • Prewet with acetone or methanol for aqueous solutions
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHydrophobic PTFE
  • Pore Size (µm).20
  • Diameter (mm)37
  • DescriptionPTFE Membrane, 0.20 37mm Wp 100/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Durable laminated PTFE membranes
  • Prewet with acetone or methanol for aqueous solutions
  • 1 year warranty
More About this Item
Membranes withstand temperatures from –100 to 130°C (–148 to 266°F). Use these hydrophobic membranes for sterile venting and filtration of gas, air steams, and organic solvents.