Cole-Parmer Essentials Precision Stainless Steel Tweezers, Tapered Pointed Ultra Fine Tips, 4-SA, 110mm
Representative image only.
Cole-Parmer Essentials Item # WZ-07387-07
Ideal for precision handling applications
  • Antimagnetic and antiacid stainless steel construction
  • Rugged and corrosion-resistant design
PRODUCT OPTIONS
£11.29GBP / Each
£13.55GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4 1/4
  • Overall Length (cm)11
  • MaterialStainless Steel Antimagnetic
  • SterileNo
  • Style4 Tapered Pointed Ultra Fine Tips
  • DescriptionPrecision Stainless Steel Tweezers, Tapered Pointed Ultra Fine Tips, 4-SA, 110mm
Key Features
  • Antimagnetic and antiacid stainless steel construction
  • Rugged and corrosion-resistant design
More About this Item
These precision stainless steel tweezers offer good corrosion resistance to most chemicals, salts, and acids. Great for lab work such as handling samples or specimens, and observing them under a microscope. The antimagnetic nature of the material makes these tweezers useful in the electronic, watch, and jewelry industries.