Cole-Parmer PolarSafe® Liquid Nitrogen Refill Tank, Stainless Steel, 30L
Representative image only.
Cole-Parmer Item # WZ-44260-68
Self-pressurized LN2 tank makes LN2 discharge safe and simple
  • Durable welded stainless steel construction ensures long-lasting performance and reduced evaporation loss
  • Unique design utilizes pressure from LN2 to discharge LN2 to other containers
  • Use as a self-pressurized LN2 backup system for cryogenic freezers
  • 5 year vacuum warranty
  • Holds a volume of up to 30 liters of liquids nitrogen
  • 1 year warranty
PRODUCT OPTIONS
£2,849.76GBP / Each
£3,419.71GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Volume (Liters)30
  • OutsideDiameter (OD) (in)17 29/32
  • Height (in)34.6
  • Evaporation (Liters/day)0.75
  • Warranty1 year
1 Year Warranty
Key Features
  • Durable welded stainless steel construction ensures long-lasting performance and reduced evaporation loss
  • Unique design utilizes pressure from LN2 to discharge LN2 to other containers
  • Use as a self-pressurized LN2 backup system for cryogenic freezers
  • 5 year vacuum warranty
  • Holds a volume of up to 30 liters of liquids nitrogen
  • 1 year warranty
More About this Item
The Cole-Parmer® PolarSafe® stainless steel liquid nitrogen (LN2) refill tanks feature a unique design ideal for cryopreservation applications. The tanks are self-pressurized utilizing pressure generated from the vaporization of small amounts of LN2 within the tank. This pressure allows for the discharge of LN2 to other containers. This unique design is more stable compared to traditional backup systems due to the reduction of intermediate pipe connections. These tanks also feature a lower working pressure, making them safer to use.