Cole-Parmer Glass Media Bottles, Class A, Round, 25 mL (0.8 oz); 10/Pk

Representative image only.
Cole-Parmer Item # WZ-34503-80
Class A round glass media storage bottles ideal for mixing, sampling, and storing mixtures, reagents, solvents, and solutions
  • Wide-mouth opening for easy pouring
  • Resistant to corrosion by most chemicals
  • Withstand temperatures from -50 to 550 °C (-58 to 1022 °F)
  • Autoclavable PP cap and bottle
  • 5 year warranty

PRODUCT OPTIONS

£17.20GBP / Pkg of 10
£20.64GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)25
  • MaterialBorosilicate Glass
  • Min Graduation (mL)5
  • Max Graduation (mL)20
  • Graduation Interval (mL)5
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size28 mm
  • Cap ColorBlue
  • Bottle ColorClear
  • AutoclavableYes
  • Withstands Low Temperature (° F)-58
  • Withstands High Temperature (° F)1022
  • Neck Inside Diameter (ID) (mm)22
  • Neck Inside Diameter (ID) (in)8 45/64
  • Outside Diameter (OD) (in)1 2/5
  • Outside Diameter (OD) (cm)3.5
  • Overall Height (in)3
  • Overall Height (cm)7.5
  • DescriptionGlass Media Bottles, Class A, Round, 25mL (0.8 oz); 10/PK
  • Withstands High Temperature (DegreeC)550
  • Withstands Low Temperature (DegreeC)-50
  • Warranty2 year
Two year warranty

Key Features

  • Wide-mouth opening for easy pouring
  • Resistant to corrosion by most chemicals
  • Withstand temperatures from -50 to 550 °C (-58 to 1022 °F)
  • Autoclavable PP cap and bottle
  • 5 year warranty

More About this Item

Made of borosilicate 3.3 glass, these Class A, round media bottles provide outstanding corrosion resistance and meet ASTM E438 requirements. Able to withstand temperatures from -50 to 550 °C (-58 to 1022 °F), they have a wide mouth opening that allows for easy pouring. The clear, colorless walls have a marking spot for labeling and graduations showing approximate volume. Both the bottle and polypropylene cap are autoclavable.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat