Cole-Parmer pH Probe, Refillable, High-temp, pH with high Na+
Representative image only.
Cole-Parmer Item # WZ-05991-32
Faster response and increased stability across a pH range of 0 to 14
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
PRODUCT OPTIONS
£126.77GBP / Each
£152.12GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsHi temp and pH with high Sodium
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)110
  • ConnectorBNC
  • DescriptionpH Probe, Refillable, High-temp, pH with high Na
Key Features
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
More About this Item
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.