Cole-Parmer pH electrode, Extra-Long, 220x6mm
Representative image only.
Cole-Parmer Item # WZ-05990-45
Combines quality and durability
  • Epoxy-body housing resists impact
  • 6 month warranty
PRODUCT OPTIONS
£191.84GBP / Each
£230.21GBP (incl VAT)
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsTest tube Sampling
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionpH electrode, Extra-Long, 220x6mm
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Epoxy-body housing resists impact
  • 6 month warranty
More About this Item
These quality laboratory electrodes have a sealed gel reference unless otherwise noted.