Cole-Parmer pH electrode, combination, double-junction, sealed, for flow-through cell 05662-40
Representative image only.
Cole-Parmer Item # WZ-05662-44
Ideal for fermentation applications
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
  • 1 year warranty
PRODUCT OPTIONS
£314.50GBP / Each
£377.40GBP (incl VAT)
Often Bought with…
Cole-Parmer pH flow-through cell, 6000 uL, 7 mm ID, glass
£163.15GBP / Each
£195.78GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • ConnectorDepends upon cable purchased separately: BNC or Ingold K9
  • DescriptionpH electrode, combination, double-junction, sealed, for flow-through cell 05662-40
  • Warranty1 year
1 Year Warranty
Key Features
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
  • 1 year warranty
More About this Item
Replacement electrodes for your flow-through cell. Select the electrode that is compatible with your model of cell, with sealed and refillable options available.