Cole-Parmer Perforated tray for models 08855-00, -02, -10
Representative image only.
Cole-Parmer Item # WZ-08855-41
Optimize performance of your ultrasonic cleaner with these accessories
  • 1 year warranty
PRODUCT OPTIONS
£331.99GBP / Each
£398.39GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPerforated tray for models 08855-00, -02, -10
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Baskets should be used to hold contents within your ultrasonic cleaner to avoid damaging the inside and bottom of your cleaner. Utilize the cover to avoid heat lose during operation and optimize temperature uniformity.