This item is now discontinued. Please browse similar items
Cole-Parmer Perforated accessory basket for 08847-00, -02 ultrasonic cleaner
Representative image only.
Cole-Parmer Item # WZ-08847-60
Optimize performance of your ultrasonic cleaner with these accessories
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPerforated accessory basket for 08847-00, -02 ultrasonic cleaner
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Baskets should be used to hold contents within your ultrasonic cleaner to avoid damaging the inside and bottom of your cleaner. Utilize the cover to avoid heat lose during operation and optimize temperature uniformity.