This item is now discontinued. Please browse similar items
Cole-Parmer Penny Head Hollow Stopper, PTFE, Size 19
Representative image only.
Cole-Parmer Item # WZ-06301-08
Snug fit and easy removal – will not freeze or bind
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number19
  • Top Outside Diameter (OD) (mm)19
  • Bottom Outside Diameter (OD) (mm)18
  • Height (mm)1
  • ColorWhite
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionPenny Head Hollow Stopper, PTFE, Size 19
More About this Item
Durable, chemically inert white PTFE stoppers are ideal for use with flasks, funnels, and cylinders that require flask-length stoppers. Hollow top minimizes expansion/contraction caused by heating or cooling. You can also use the hollow top as a container for micro-analytical processing of solutions.